Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIF1 alpha Antibody (PCRP-TRIM24-1B12), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB378721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378721 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378721 Supplier Novus Biologicals Supplier No. NBP308786IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIF1 alpha Monoclonal antibody specifically detects TIF1 alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen TIF1 alpha
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-TRIM24-1B12
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24
Host Species Mouse
Immunogen Recombinant full-length human TIF1 alpha protein (Uniprot: O15164)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 8805
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.