Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGAR/C12orf5 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP149534W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149534W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149534W Supplier Novus Biologicals Supplier No. NBP149534AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIGAR/C12orf5 Polyclonal specifically detects TIGAR/C12orf5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TIGAR/C12orf5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9NQ88
Gene Alias chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein
Gene Symbols TIGAR
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57103
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.