Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGAR/C12orf5 Antibody (M2-P4H2), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB212478 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212478 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212478 Supplier Novus Biologicals Supplier No. NB1002698DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIGAR/C12orf5 Monoclonal antibody specifically detects TIGAR/C12orf5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TIGAR/C12orf5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M2-P4H2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein
Host Species Mouse
Immunogen Synthetic peptide; IINFEEGREV (amino acids 239-248)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Biologically Active Proteins, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57103
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.