Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGAR/C12orf5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224754 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224754 0.1 mg
NBP224754SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224754 Supplier Novus Biologicals Supplier No. NBP224754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIGAR/C12orf5 Polyclonal specifically detects TIGAR/C12orf5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TIGAR/C12orf5
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-4 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Accession No. Q9NQ88
Gene Alias chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein
Gene Symbols TIGAR
Host Species Rabbit
Immunogen A portion of amino acids 140-190 of human TIGAR was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57103
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human and 93% homologous in chimp.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.