Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGAR/C12orf5 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$160.07 - $406.62

Specifications

Antigen TIGAR/C12orf5
Concentration 0.5 mg/mL
Dilution Western Blot 1-4 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224754SS
SDP
View Documents
Novus Biologicals
NBP224754SS
0.025 mg
Each for $160.07
Only null left
Add to Cart
 
NBP224754
SDP
View Documents
Novus Biologicals
NBP224754
0.1 mg
Each for $406.62
Only null left
Add to Cart
 
Description

Description

TIGAR/C12orf5 Polyclonal specifically detects TIGAR/C12orf5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TIGAR/C12orf5
Western Blot 1-4 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat
Q9NQ88
57103
A portion of amino acids 140-190 of human TIGAR was used as the immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Lipid and Metabolism
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein
TIGAR
IgG
Protein G purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human and 93% homologous in chimp.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.