Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGD1 Antibody (OTI4G9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP274519G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274519G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274519G Supplier Novus Biologicals Supplier No. NBP274519G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIGD1 Monoclonal specifically detects TIGD1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TIGD1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4G9
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EEYORE, tigger transposable element derived 1, tigger transposable element-derived protein 1
Gene Symbols TIGD1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-267 of human TIGD1 (NP_663748) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 200765.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.