Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tight Junction Protein 1 Antibody (HL1185) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294843 Supplier Novus Biologicals Supplier No. NBP325774
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tight Junction Protein 1 Monoclonal antibody specifically detects Tight Junction Protein 1 in Human, Mouse, Rat, Canine, Feline samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Tight Junction Protein 1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HL1185
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence 1:100-1:1000
Formulation PBS
Gene Alias DKFZp686M05161, MGC133289, Tight junction protein 1, tight junction protein 1 (zona occludens 1), tight junction protein ZO-1, TJP1, ZO1, ZO-1, zona occludens 1, Zona occludens protein 1, zonula occludens 1 protein, Zonula occludens protein 1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Tight Junction Protein 1. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7082
Target Species Human, Mouse, Rat, Canine, Feline
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.