Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tight Junction Protein 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018653 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018653 0.025 mg
NBP14966905 0.05 mg
NBP14966901 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018653 Supplier Novus Biologicals Supplier No. NBP1496690.025MG
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody has been used in 2 publications

Tight Junction Protein 1 Polyclonal specifically detects Tight Junction Protein 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Tight Junction Protein 1
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS, containing 0.1% BSA with 0.02% Sodium Azide
Gene Accession No. Q07157
Gene Alias DKFZp686M05161, MGC133289, Tight junction protein 1, tight junction protein 1 (zona occludens 1), tight junction protein ZO-1, TJP1, ZO1, ZO-1, zona occludens 1, Zona occludens protein 1, zonula occludens 1 protein, Zonula occludens protein 1
Gene Symbols TJP1
Host Species Guinea Pig
Immunogen The polyclonal antibody is raised against a human peptide antigen corresponding to ten residues at the splice junction and as such recognizes specifically ZO-1 alpha-minus.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7082
Test Specificity The polyclonal antibody is raised against a human peptide antigen corresponding to ten residues at the splice junction and as such recognizes specifically ZO-1 alpha-minus.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.