Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tight Junction Protein 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB119909 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119909 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119909 Supplier Novus Biologicals Supplier No. NBP311284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tight Junction Protein 1 Polyclonal specifically detects Tight Junction Protein 1 in Human, Mouse, Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Tight Junction Protein 1
Applications Flow Cytometry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias DKFZp686M05161, MGC133289, Tight junction protein 1, tight junction protein 1 (zona occludens 1), tight junction protein ZO-1, TJP1, ZO1, ZO-1, zona occludens 1, Zona occludens protein 1, zonula occludens 1 protein, Zonula occludens protein 1
Host Species Rabbit
Immunogen Recombinant human GST-Tight Junction Protein 1 (aa 1-888)
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7082
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.