Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tight Junction Protein 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB018652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018652 0.025 mg
NBP14966705 0.05 mg
NBP14966701 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018652 Supplier Novus Biologicals Supplier No. NBP1496670.025MG
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Tight Junction Protein 2 Polyclonal specifically detects Tight Junction Protein 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Tight Junction Protein 2
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation 0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide
Gene Alias C9DUPq21.11, DFNA51, DUP9q21.11, MGC26306, Tight junction protein 2, tight junction protein 2 (zona occludens 2), TJP2, X104, X104tight junction protein ZO-2, ZO2, ZO-2, ZO2Friedreich ataxia region gene X104 (tight junction protein ZO-2), Zona occludens protein 2, Zonula occludens protein 2
Gene Symbols TJP2
Host Species Guinea Pig
Immunogen For immunisation of the guinea pig, amino acids 411-467 of human ZO-2 were used as immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9414
Test Specificity ZO-2 can interact with zona occludens 1 (ZO-1). Furthermore, the PDZ2 domain of ZO-2 was shown to interact with connexin-43, the predominant connexin in epithelial and most other tissues which is involved in cell growth control and embryonic development. The antibody cross reacts with mouse ZO-2.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.