Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tight Junction Protein 2 Rabbit anti-Human, Biotin, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB056394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056394 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056394 Supplier Novus Biologicals Supplier No. NBP298900B

Rabbit Polyclonal Antibody

Tight Junction Protein 2 Polyclonal specifically detects Tight Junction Protein 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Tight Junction Protein 2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C9DUPq21.11, DFNA51, DUP9q21.11, MGC26306, Tight junction protein 2, tight junction protein 2 (zona occludens 2), TJP2, X104, X104tight junction protein ZO-2, ZO2, ZO-2, ZO2Friedreich ataxia region gene X104 (tight junction protein ZO-2), Zona occludens protein 2, Zonula occludens protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Tight Junction Protein 2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9414
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.