Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT Antibody (2190A), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB311308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311308 Supplier Novus Biologicals Supplier No. FAB72671APCCY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TIGIT Monoclonal antibody specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen TIGIT
Applications Flow Cytometry
Classification Monoclonal
Clone 2190A
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TIGIT, Met1-Thr143, Accession # P86176
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 201633
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.