Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT Antibody (2629A) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB401436 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401436 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401436 Supplier Novus Biologicals Supplier No. FAB10532I
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TIGIT Monoclonal antibody specifically detects TIGIT in Cynomolgus Monkey samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF

Specifications

Antigen TIGIT
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Classification Monoclonal
Clone 2629A
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM
Host Species Rabbit
Immunogen Human embryonic kidney cell HEK293-derived Cynomolgus monkey TIGIT, Met22-Pro142, Accession # XP_00554815
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 201633
Target Species Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.