Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT Antibody (TIGIT/3106), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB012764 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012764 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012764 Supplier Novus Biologicals Supplier No. NBP279928AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Product Image.

Specifications

Antigen TIGIT
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TIGIT/3106
Conjugate Alexa Fluor 647
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM
Gene Symbols TIGIT
Host Species Mouse
Immunogen Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary) (Uniprot: Q4951)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.