Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIGIT Recombinant Rabbit Monoclonal Antibody (BLR047F)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | TIGIT |
---|---|
Clone | BLR047F |
Concentration | 1 mg/mL |
Content And Storage | 2°C to 8°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544289
|
Invitrogen™
MA544289 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544290
|
Invitrogen™
MA544290 |
10 μL |
Each for $173.10
|
|
|||||
Description
TIGIT Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.Specifications
TIGIT | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
Q495A1 | |
201633 | |
A peptide representing a region between residue 194 and the C-terminus (residue 244) of human T-cell immunoreceptor with Ig and ITIM domains precursor (TIGIT). | |
Antibody |
BLR047F | |
2°C to 8°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
TIGIT | |
ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
TIGIT | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title