Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-1/KIM-1/HAVCR Antibody (104), FITC
SDP

Catalog No. NB078582 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB078582 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB078582 Supplier Novus Biologicals Supplier No. NBP290831F

Rabbit Monoclonal Antibody

TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Rat samples. It is validated for ELISA.

Specifications

Antigen TIM-1/KIM-1/HAVCR
Applications ELISA
Classification Monoclonal
Clone 104
Conjugate FITC
Formulation PBS
Gene Alias HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat TIM-1/KIM-1/HAVCR (aa18-238; NP_775172.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26762
Test Specificity No cross-reactivity in ELISA with: Mouse KIM-1/ TIM1 Human KIM-1/ TIM1 Human TIMD3
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.