Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-1/KIM-1/HAVCR Antibody (990605), HRP, Novus Biologicals™
SDP

Catalog No. NB349682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349682 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349682 Supplier Novus Biologicals Supplier No. FAB17503H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TIM-1/KIM-1/HAVCR
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 990605
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human TIM-1/KIM-1/HAVCR, Met1-Thr288, Accession # Q96D42
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Cellular Signaling, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 26762
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.