Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-1/KIM-1/HAVCR Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 19 publications

$244.50 - $584.50

Specifications

Antigen TIM-1/KIM-1/HAVCR
Concentration 1 mg/mL
Dilution Western Blot 1-8 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml, KnockDown Validated
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP176701SS
SDP
View Documents
Novus Biologicals
NBP176701SS
0.025 mg
Each for $244.50
Only null left
Add to Cart
 
NBP176701
SDP
View Documents
Novus Biologicals
NBP176701
0.1 mg
Each for $584.50
Only null left
Add to Cart
 
Description

Description

TIM-1/KIM-1/HAVCR Polyclonal specifically detects TIM-1/KIM-1/HAVCR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

TIM-1/KIM-1/HAVCR
Western Blot 1-8 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml, KnockDown Validated
Polyclonal
Rabbit
Human, Mouse, Rat
Q96D42
26762
Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human TIM-1. The immunogen is located within amino acids 50 - 100 of TIM-1. Amino Acid Squence: TNGTHVTYRKDTRYKL
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
44 kDa
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS with 0.02% Sodium Azide
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
HAVCR1
IgG
Affinity Purified
Human TIM-1/KIM-1/HAVCR has 5 isoforms, including isoform a precursor (364aa, 39kD), isoform a (364aa, 39kD), isoform b precursor (401aa, 44kD), isoform b (359aa, 39kD), and isoform c (359aa, 39kD). This antibody detects human isoform b precursor. Mouse TIM-1/KIM-1/HAVCR has 3 isoforms, including isoform a precursor (305aa, 33kD), isoform b precursor (282aa, 31kD) and isoform b (282aa, 31kD). Rat TIM-1/KIM-1/HAVCR has only one isoform identified so far (307aa, 34kD).
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.