Learn More
Description
Specifications
Specifications
| Antigen | TIM-1/KIM-1/HAVCR |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | KIM70 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, ELISA, Sandwich ELISA |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 |
| Gene Symbols | HAVCR1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
