Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-1/KIM-1/HAVCR Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB354982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354982 Supplier Novus Biologicals Supplier No. NB10056421MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIM-1/KIM-1/HAVCR Polyclonal specifically detects TIM-1/KIM-1/HAVCR in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen TIM-1/KIM-1/HAVCR
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Host Species Rabbit
Immunogen A synthetic peptide (SPLYSYTTDGNDTVT) of human TIM-1 protein was used as immunogen for this antibody. This sequence corresponds to amino acids (aa) 248-262 of NP_036338 and aa 84-98 of EAW61615.1. Isoforms of TIM-1 have been described. For example, NP_036338 is 364 aa and EAW61615.1 is 194 aa. EAW61615.1 is referred to as isoform CRA_a.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Cellular Signaling, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 26762
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.