Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 Antibody (2321C), Biotin, Novus Biologicals™
SDP

Catalog No. NB365472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365472 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365472 Supplier Novus Biologicals Supplier No. FAB23652B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen TIM-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 2321C
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIM-3, Met1-Arg200, Accession # Q8TDQ0
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84868
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.