Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 Antibody (F38-2E2) - Azide Free, Novus Biologicals™
SDP

Catalog No. NBP227221 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227221 0.1 mg
NBP227221SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227221 Supplier Novus Biologicals Supplier No. NBP227221
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface), CyTOF-ready.

Specifications

Antigen TIM-3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone F38-2E2
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Flow (Cell Surface), CyTOF-ready
Gene Accession No. Q8TDQ0
Gene Alias CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Gene Symbols HAVCR2
Host Species Mouse
Immunogen This antibody was raised against recombinant human TIM-3. [Uniprot# Q8TDQ0]
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84868
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.