Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 Antibody (OTI2E2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27420A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27420A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27420A32 Supplier Novus Biologicals Supplier No. NBP274520AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen TIM-3
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2E2
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Gene Symbols HAVCR2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 22-202 of human TIM-3 (NP_116171) produced in E.coli.
Quantity 0.1 mL
Research Discipline Cancer, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84868
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.