Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-4 Antibody (1018144), HRP, Novus Biologicals™
SDP

Catalog No. NB348711 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348711 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348711 Supplier Novus Biologicals Supplier No. FAB2826H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TIM-4 Monoclonal specifically detects TIM-4 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TIM-4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1018144
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4
Host Species Rat
Immunogen Mouse myeloma cell line, NS0-derived mouse TIM-4, Ala22-Thr279, Accession # Q6U7R4
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 91937
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.