Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-4 Antibody (102), Alexa Fluor™ 647
SDP

Catalog No. NB074923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074923 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074923 Supplier Novus Biologicals Supplier No. NBP290556AF647

Rabbit Monoclonal Antibody

TIM-4 Monoclonal specifically detects TIM-4 in Mouse samples. It is validated for ELISA.

Specifications

Antigen TIM-4
Applications ELISA
Classification Monoclonal
Clone 102
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TIM-4 (NP_848874.3; Met 1-Thr 279).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91937
Test Specificity No cross-reactivity in ELISA with: Human TIMD4 Human KIM1 (aa 1-135)
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.