Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-4 Antibody (921832), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB363723 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363723 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363723 Supplier Novus Biologicals Supplier No. FAB2929L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIM-4 Monoclonal antibody specifically detects TIM-4 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen TIM-4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 921832
Conjugate DyLight 550
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4
Host Species Mouse
Immunogen Mouse myeloma cell line NSO-derived recombinant human TIM-4, Glu25-Leu315, Accession # Q96H15
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 91937
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.