Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIM3 Recombinant Rabbit Monoclonal Antibody (BLR033F)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | TIM3 |
---|---|
Clone | BLR033F |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544261
|
Invitrogen™
MA544261 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544262
|
Invitrogen™
MA544262 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
TIM3 (Hepatitis A virus cellular receptor 2, HAVCR2, T-cell immunoglobulin, mucin-dmain containing-3) is a 281 amino acid long, Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. TIM3 consists of an IgV-like domain, a mucin-like domain in the extracellular region, and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. TIM3 is involved in macrophage activation and induction of autoimmune diseases. Further, TIM3 down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of TIM3 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease, and also enhances the severity of experimental autoimmune encephalomyelitis in mice.Specifications
TIM3 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
Q8TDQ0 | |
84868 | |
Recombinant protein representing the extracellular domain of human TIM3 (HAVCR2) (residues 22-201). | |
Antibody |
BLR033F | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
Havcr2 | |
CD366; FLJ14428; Havcr2; HAVcr-2; Hepatitis A virus cellular receptor 2; hepatitis A virus cellular receptor 2 homolog; kidney injury molecule-3; KIM-3; sCD366; soluble CD366; soluble TIM 3; T cell immunoglobulin mucin 3; T cell immunoglobulin mucin-3; T-cell immunoglobulin and mucin domain containing 3; T-cell immunoglobulin and mucin domain-containing protein 3; T-cell immunoglobulin mucin family member 3; T-cell immunoglobulin mucin receptor 3; T-cell membrane protein 3; Tim3; TIM-3; TIMD3; TIMD-3 | |
Havcr2 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title