Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMBIM 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB11087040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11087040 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11087040 Supplier Novus Biologicals Supplier No. NB11087040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIMBIM 1 Polyclonal specifically detects TIMBIM 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen TIMBIM 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 10ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Dot Blot
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q969X1
Gene Alias TMBIM 1, TMBIM1, Transmembrane BAX inhibitor motif containing 1, Transmembrane BAX inhibitor motif containing protein 1
Host Species Rabbit
Immunogen This protein A purified TIMBIM 1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to the amino terminus of human TIMBIM 1 protein. (Uniprot: Q969X1)
Molecular Weight of Antigen 34.61 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is specific for human TMBIM1 protein.
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.