Learn More
Invitrogen™ TIMELESS Polyclonal Antibody

Description
Immunogen sequence: FVELLFWKNT AVVREMTEGY GSLDDRSSSR RAPTWSPEEE AHLQELYLAN KDVEGQDVVE AILAHLNTVP RTRKQIIHHL VQMGLADSVK DFQR Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
Specifications
Specifications
| Antigen | TIMELESS |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TIMELESS |
| Gene Accession No. | Q9UNS1 |
| Gene Alias | C77407; Debt69; hTIM; mTim; Protein timeless homolog; RGD:620939}; rTIM; rTLP; TIM; Tim1; timeless; timeless {ECO:0000312; timeless circadian clock; timeless circadian clock 1; timeless circadian regulator; timeless homolog; TIMELESS1; timeless-like protein; Tof1 homolog; unknown |
| Gene Symbols | TIMELESS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.