Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Timeless Rabbit anti-Human, Clone: 6D4L2, Novus Biologicals™
SDP

Catalog No. NB165641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165641 20 μg
NB165640 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165641 Supplier Novus Biologicals Supplier No. NBP31527220UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Timeless Monoclonal antibody specifically detects Timeless in Human samples. It is validated for Western Blot

Specifications

Antigen Timeless
Applications Western Blot
Classification Monoclonal
Clone 6D4L2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ12640, hTIMTIMELESS1, TIM1protein timeless homolog, timeless (Drosophila) homolog, timeless homolog (Drosophila), TIMFLJ20714
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1058-1208 of human Timeless (Q9UNS1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 8914
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.