Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM17A Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB045066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045066 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045066 Supplier Novus Biologicals Supplier No. NBP297571R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIMM17A Polyclonal specifically detects TIMM17A in Human samples. It is validated for Western Blot.

Specifications

Antigen TIMM17A
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Inner membrane preprotein translocase Tim17a, mitochondrial import inner membrane translocase subunit Tim17-A, mitochondrial inner membrane translocase, TIM17Apreprotein translocase, TIM17MIMT17, TIMM17, translocase of inner mitochondrial membrane 17 homolog A (yeast)
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TIMM17A fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10440
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.