Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM50 Antibody (SR1606), Novus Biologicals™
SDP

Catalog No. NB387902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387902 100 μg
NB387901 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387902 Supplier Novus Biologicals Supplier No. NBP322096100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TIMM50 Monoclonal antibody specifically detects TIMM50 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen TIMM50
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR1606
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Formulation PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Gene Alias homolog of yeast Tim50, MGC102733, mitochondrial import inner membrane translocase subunit TIM50, TIM50, TIM50L, Tim50-like protein, translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae), translocase of inner mitochondrial membrane 50 homolog (yeast)
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIMM50 (Uniprot #: Q3ZCQ8)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 92609
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.