Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM8A Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB051360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051360 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051360 Supplier Novus Biologicals Supplier No. NBP298033C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIMM8A Polyclonal specifically detects TIMM8A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TIMM8A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DDP1deafness/dystonia peptide, DDPMGC12262, Deafness dystonia protein 1, DFN1, mitochondrial import inner membrane translocase subunit Tim8 A, MTSTIM8, TIM8A, translocase of inner mitochondrial membrane 8 (yeast) homolog A, translocase of inner mitochondrial membrane 8 homolog A (yeast), X-linked deafness dystonia protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TIMM8A fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1678
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.