Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-1 Antibody (63515), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB302357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302357 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302357 Supplier Novus Biologicals Supplier No. FAB970R

Mouse Monoclonal Antibody

TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen TIMP-1
Applications Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 63515
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIMP-1, Cys24-Ala207, Accession # P01033
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 7076
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.