Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-1 Antibody (TIMP1/1944R), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB384910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384910 Supplier Novus Biologicals Supplier No. NBP308713AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human samples. It is validated for ELISA

Specifications

Antigen TIMP-1
Applications ELISA
Classification Monoclonal
Clone TIMP1/1944R
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
Host Species Rabbit
Immunogen Recombinant human full-length TIMP-1 protein (Uniprot: P01033)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 7076
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.