Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-4 Antibody (9:4-7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB151266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151266 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151266 Supplier Novus Biologicals Supplier No. NBP221658AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIMP-4 Monoclonal specifically detects TIMP-4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TIMP-4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9:4-7
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4
Host Species Mouse
Immunogen TIMP-4 Antibody (9:4-7) was produced in mouse by repeated immunizations with full length human TIMP-4 protein followed by hybridoma development. (Uniprot: Q99727)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7079
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.