Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-4 Antibody (MM0037-9F3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB11061003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11061003 0.1 mg
NB017995 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11061003 Supplier Novus Biologicals Supplier No. NB11061003
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIMP-4 Monoclonal specifically detects TIMP-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TIMP-4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0037-9F3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:1000, Immunohistochemistry 1:100-200, Immunohistochemistry-Paraffin 1:100-1:200
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Alias metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4
Gene Symbols TIMP4
Host Species Mouse
Immunogen Human recombinant TIMP-4
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7079
Test Specificity No cross-reactivity to human TIMP-1, 2 and 3 is noted
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.