Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-4 Antibody (MM0037-9F3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB11061003G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11061003G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11061003G Supplier Novus Biologicals Supplier No. NB11061003G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIMP-4 Monoclonal specifically detects TIMP-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TIMP-4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0037-9F3
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4
Gene Symbols TIMP4
Host Species Mouse
Immunogen Human recombinant TIMP-4
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cellular Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7079
Test Specificity No cross-reactivity to human TIMP-1, 2 and 3 is noted
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.