Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tiotidine
GREENER_CHOICE

Catalog No. p-4785680
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 08-261-0

Supplier: Tocris Bioscience™ 0826/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective H2 antagonist

Potent histamine H2-receptor antagonist with negligible activity against H1- and H3- receptors.

Chemical Identifiers

CAS 69014-14-8
Molecular Formula C10H16N8S2

Specifications

Quantity 10 mg
Formula Weight 312.41
Purity >99%
Molecular Weight (g/mol) 312.414
Chemical Name or Material Tiotidine
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.