Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIP30 Antibody (5D6), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB162275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162275 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162275 Supplier Novus Biologicals Supplier No. NBP259482G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TIP30 Monoclonal antibody specifically detects TIP30 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen TIP30
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 5D6
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias 30 kDa HIV-1 TAT-interacting protein, CC3oxidoreductase HTATIP2, EC 1.1.1, EC 1.1.1.-, FLJ26963, HIV-1 Tat interactive protein 2, 30 kDa, HIV-1 Tat interactive protein 2, 30kDa, HIV-1 TAT-interactive protein 2, SDR44U1, Tat-interacting protein (30kD), TIP30short chain dehydrogenase/reductase family 44U, member 1
Host Species Mouse
Immunogen Recombinant human TIP30 (1-242aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10553
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.