Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIP30 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB054227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054227 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054227 Supplier Novus Biologicals Supplier No. NBP298510F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIP30 Polyclonal specifically detects TIP30 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TIP30
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 30 kDa HIV-1 TAT-interacting protein, CC3oxidoreductase HTATIP2, EC 1.1.1, EC 1.1.1.-, FLJ26963, HIV-1 Tat interactive protein 2, 30 kDa, HIV-1 Tat interactive protein 2, 30kDa, HIV-1 TAT-interactive protein 2, SDR44U1, Tat-interacting protein (30kD), TIP30short chain dehydrogenase/reductase family 44U, member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TIP30 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10553
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.