Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIP60 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220647 Supplier Novus Biologicals Supplier No. NBP220647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIP60 Polyclonal specifically detects TIP60 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TIP60
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.01mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Accession No. Q92993
Gene Alias cPLA2, ESA1EC 2.3.1.48, Histone acetyltransferase HTATIP, histone acetyltransferase KAT5, HIV-1 Tat interactive protein, HIV-1 Tat interactive protein, 60kDa, HTATIP1, HTATIPcPLA2 interacting protein, K(lysine) acetyltransferase 5,60 kDa Tat-interactive protein, K-acetyltransferase 5, Lysine acetyltransferase 5, PLIPTIP, Tat interacting protein, 60kDa, Tip60, TIP60cPLA(2)-interacting protein
Gene Symbols KAT5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TIP60. The exact sequence is proprietary.
Molecular Weight of Antigen 59 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 10524
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.