Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIPIN Antibody (OTI2E6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N274528A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274528A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274528A405 Supplier Novus Biologicals Supplier No. NBP274528AF405

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

TIPIN Monoclonal specifically detects TIPIN in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TIPIN
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2E6
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ20516, TIMELESS interacting protein, TIMELESS-interacting protein
Gene Symbols TIPIN
Host Species Mouse
Immunogen Full length recombinant protein of human TIPIN (NP_060328) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 54962.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.