Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIPRL Antibody (OTI3D11), FITC, Novus Biologicals™
SDP

Catalog No. NBP274529F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274529F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274529F Supplier Novus Biologicals Supplier No. NBP274529F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIPRL Monoclonal specifically detects TIPRL in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen TIPRL
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3D11
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias dJ69E11.3, MGC3794, putative MAPK activating protein, Putative MAPK-activating protein PM10, TIP41, TOR signaling pathway regulator-like (S. cerevisiae), TIP41-like protein, TIP41Type 2A-interacting protein, TIPTIP41, TOR signalling pathway regulator-like (S. cerevisiae)
Gene Symbols TIPRL
Host Species Mouse
Immunogen Full length recombinant protein of human TIPRL (NP_690866) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 261726.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.