Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
SDP

For use in research applications

$706.00 - $1452.00

Specifications

Host Species Human, Mouse
Components TIRAP Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKLQLRDAAPGGAIVS (TIRAP sequence is underlined). Molecular weight: 3701.4. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361.
For Use With (Application) Inhibition of TIRAP binding to TLR2 or TLR4
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Product Type TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP229331
SDP
View Documents
Novus Biologicals™
NBP229331
2 mg
Each for $706.00
Only null left
Add to Cart
 
NBP2293315
SDP
View Documents
Novus Biologicals™
NBP2293315MG
5 mg
Each for $1,452.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Human, Mouse
Inhibition of TIRAP binding to TLR2 or TLR4
TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
TLR2, TLR4
TIRAP Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKLQLRDAAPGGAIVS (TIRAP sequence is underlined). Molecular weight: 3701.4. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361.
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
3701.4
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.