Learn More
Novus Biologicals™ TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
SDP
Applications: Flow Cytometry, In vitro assay
$706.00
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP226245
SDP
View Documents
Novus Biologicals™
NBP226245
1 mg
Each of 1 for $706.00
Only null left
Add to Cart
Description
Description
TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.