Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tissue alpha-L-Fucosidase/FUCA1 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB064882 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064882 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064882 Supplier Novus Biologicals Supplier No. NBP300209DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tissue alpha-L-Fucosidase/FUCA1 Polyclonal specifically detects Tissue alpha-L-Fucosidase/FUCA1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Tissue alpha-L-Fucosidase/FUCA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Alpha-L-fucosidase 1, Alpha-L-fucosidase I, Alpha-L-fucoside fucohydrolase 1, EC 3.2.1, EC 3.2.1.51, FUCA, fucosidase, alpha-L- 1, tissue, tissue alpha-L-fucosidase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Tissue alpha-L-Fucosidase/FUCA1 (P04066; Gln32-Lys466).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2517
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.