Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TissuePrep™ Embedding Media (Certified), Fisher Chemical

Catalog No. t565 Shop All Fisherbrand Products
Click to view available options
Quantity:
5 lb.

Catalog No. T56-5

Chemical Identifiers

CAS 8002-74-2
Molecular Formula C21H27NO3
Molecular Weight (g/mol) 341.451
MDL Number MFCD00212755
InChI Key JWHAUXFOSRPERK-UHFFFAOYSA-N
Synonym propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn
PubChem CID 4932
ChEBI CHEBI:63619
IUPAC Name 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one
SMILES CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O

Specifications

Physical Form Solid
Melting Point 55°C to 57°C
Boiling Point 322°C
Packaging Carton
Color White
Quantity 5 lb.
Grade Certified
Chemical Name or Material TissuePrep™ Embedding Media
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.