Learn More
Titanium aluminide, -325 mesh, 99.5% (metals basis)
Description
It finds use in several applications including automobiles and aircraft. Also used in air frames, investment casting, turbocharger rotors, and wire/fiber metal matrix composites
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 10 g |
| UN Number | UN3089 |
| Solubility Information | Insoluble in water. |
| Percent Purity | 99.5% |
| Odor | Odorless |
| Sensitivity | Air Sensitive |
| Assay | (metals basis) |
| Chemical Name or Material | Titanium aluminide |
Safety and Handling
| Hazard Category | H228-H315-H319-H335 |
| Hazard Statement | GHS H Statement H228-H315-H319-H335 Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.