Alfa Aesar™ Titanium(IV) 2-ethylhexanoate, 97%

Manufacturer: Alfa Aesar™ 4460236

Quantity 500g

 View more versions of this product

Catalog No. AA4460236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3645-34-9
    Molecular Formula C32H60O8Ti
    Formula Weight 623.75
    MDL Number MFCD05663781
    Chemical Name or Material Titanium (IV) 2-ethylhexanoate

    Specifications

    Formula Weight 623.75
    Molecular Formula C32H60O8Ti
    Quantity 500g
    MDL Number MFCD05663781
    CAS 3645-34-9
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.
    TSCA TSCA
    Health Hazard 3 GHS P Statement
    P280h
    Wear protective gloves/clothing.
    Physical Form Liquid
    Odor Odorless
    Chemical Name or Material Titanium (IV) 2-ethylhexanoate
    Assay 97%

    GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P280h
    Wear protective gloves/clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit